missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC5 Antibody (CL8121) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88913MFV500
This item is not returnable.
View return policy
Description
HDAC5 Monoclonal antibody specifically detects HDAC5 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| HDAC5 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| Antigen NY-CO-9, EC 3.5.1.98, FLJ90614, HD5, histone deacetylase 5, KIAA0600, NY-CO-9 | |
| This antibody was developed using a synthetic peptide derived from Q9UQL6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific | |
| 10014 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL8121 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction