missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC4 Recombinant Rabbit Monoclonal Antibody (23GB865), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | HDAC4 |
|---|---|
| Clone | 23GB865 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089650
|
Invitrogen™
MA554010 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089005
|
Invitrogen™
MA554011 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC4 Recombinant Monoclonal antibody specifically detects HDAC4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| HDAC4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P56524 | |
| 9759 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB865 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4932408F19Rik; AHO3; AI047285; BDMR; DKFZp779K1053; HA6116; HD4; HD7; HD7b; HD9; HDAC; HDAC4; HDAC-4; HDAC7; HDAC7B; HDAC9; HDAC9B; HDAC9FL; HDACA; HDAC-A; HDRP; Histone deacetylase 4; histone deacetylase A; KIAA0288; KIAA0744; MITR | |
| A synthetic peptide of human HDAC4 (30-100AA). | |
| Primary | |
| -20°C | |
| HDAC4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title