missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC4, Mouse anti-Human, Clone: 2B11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009759-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: HAQAGVQVKQEPIESDEEEAEPPREVEPGQRQPSEQELLFRQQALLLEQQRIHQLRNYQASMEAAGIPVSFGGHRPLSRAQSSPASATFPVSVQEPPTSpecifications
| HDAC4 | |
| Monoclonal | |
| Unconjugated | |
| histone deacetylase 4 | |
| HA6116/HD4/HDAC-A/HDACA/KIAA0288 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2B11 | |
| In 1x PBS, pH 7.4 | |
| NM_006037 | |
| HDAC4 | |
| HDAC4 (NP_006028.1, 551 a.a. ∼ 648 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9759 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction