missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC4 Monoclonal Antibody (7E2E6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538538
This item is not returnable.
View return policy
Description
This antibody has been tested in indirect ELISA.
Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
Specifications
| HDAC4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P56524, Q6NZM9 | |
| HDAC4 | |
| Purified recombinant fragment of human HDAC4 (aa: 456-592) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 7E2E6 | |
| Unconjugated | |
| HDAC4 | |
| 4932408F19Rik; AHO3; AI047285; BDMR; DKFZp779K1053; HA6116; HD4; HD7; HD7b; HD9; HDAC; HDAC4; HDAC-4; HDAC7; HDAC7B; HDAC9; HDAC9B; HDAC9FL; HDACA; HDAC-A; HDRP; Histone deacetylase 4; histone deacetylase A; KIAA0288; KIAA0744; MITR | |
| Mouse | |
| Protein G | |
| RUO | |
| 208727, 9759 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction