missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC3 Mouse anti-Human, Mouse, Rat, Clone: 3E8F10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67151-1-IG-20UL
This item is not returnable.
View return policy
Description
HDAC3 Monoclonal antibody specifically detects HDAC3 in Human, Mouse, Rat samples. It is validated for WB, ELISASpecifications
| HDAC3 | |
| Monoclonal | |
| 2000 μg/mL | |
| PBS with 0.1% sodium azide and 50% glycerol pH 7.3. | |
| Mouse | |
| 20 μL | |
| 8841 | |
| Store at -20°C | |
| Liquid |
| Western Blot, ELISA | |
| 3E8F10 | |
| Unconjugated | |
| NM_001031717 | |
| Protein A purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction