Learn More
Invitrogen™ HDAC3 Monoclonal Antibody (7G6C5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515442
Description
MA5-15442 targets HDAC3 in indirect ELISA, IF, ChIP and WB applications and shows reactivity with Human samples. The MA5-15442 immunogen is purified recombinant fragment of human HDAC3 (aa224-428) expressed in E. Coli. MA5-15442 detects HDAC3 which has a predicted molecular weight of approximately 48.8kDa.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.
Specifications
| HDAC3 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| O15379 | |
| HDAC3 | |
| Purified recombinant fragment of human HDAC3 (aa224-428) expressed in E. Coli. | |
| RUO | |
| 8841 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, ELISA, Western Blot, Immunocytochemistry | |
| 7G6C5 | |
| Unconjugated | |
| HDAC3 | |
| AW537363; HD3; Hdac3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; histone deacetylase 3; histone deacetylase 3-like protein; RPD3; RPD3-2; SMAP45 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.