missing translation for 'onlineSavingsMsg'
Learn More

HDAC3 Antibody (PCRP-HDAC3-3C9), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-14061MFV450

Product Code. 18551745

  • £558.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HDAC3 Monoclonal antibody specifically detects HDAC3 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDAC3
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Flow Cytometry
PCRP-HDAC3-3C9
Flow Cytometry
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Recombinant full-length human HDAC3 protein (Uniprot: O15379)
0.1 mL
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics
8841
Store at 4C in the dark.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.