missing translation for 'onlineSavingsMsg'
Learn More

HDAC3 Antibody (PCRP-HDAC3-3C9), DyLight 650, Novus Biologicals™

Product Code. 18369664 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369664 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369664 Supplier Novus Biologicals Supplier No. NBP314061C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HDAC3 Monoclonal antibody specifically detects HDAC3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Spécification

Antigen HDAC3
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-HDAC3-3C9
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Host Species Mouse
Immunogen Recombinant full-length human HDAC3 protein (Uniprot: O15379)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 8841
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.