missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HDAC3 Monoclonal antibody specifically detects HDAC3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | HDAC3 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-HDAC3-3C9 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human HDAC3 protein (Uniprot: O15379) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?