missing translation for 'onlineSavingsMsg'
Learn More

HDAC3 Antibody (PCRP-HDAC3-2D4), DyLight 594, Novus Biologicals™

Product Code. 18360813 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360813 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360813 Supplier Novus Biologicals Supplier No. NBP314060DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HDAC3 Monoclonal antibody specifically detects HDAC3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC3
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-HDAC3-2D4
Conjugate DyLight 594
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Host Species Mouse
Immunogen Recombinant full-length human HDAC3 protein (Uniprot: O15379)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 8841
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.