missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC2 Mouse anti-Human, Unconjugated, Clone: 1A3E4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HDAC2 |
|---|---|
| Clone | 1A3E4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16849645
|
Proteintech
67165-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16839645
|
Proteintech
67165-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HDAC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC2. This antibody recognizes human, mouse, rat antigen. The HDAC2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| HDAC2 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 3066 | |
| Primary |
| 1A3E4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title