Learn More
Invitrogen™ HDAC11 Recombinant Rabbit Monoclonal Antibody (9I4N2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557878
Description
Immunogen sequence: MLHTTQLYQH VPETRWPIVY SPRYNITFMG LEKLHPFDAG KWGKVINFLK EEKLLSDSML VEAREASEED LLVVHTRRYL NELKWSFAVA TITEIPPVIF LPNFLVQRKV LRPLRTQTGG TIMAGKLAVE RGWAINVGGG FHHCSSDRGG GFCAYADITL AIKFLFERVE GISRATIIDL DAHQGNGHER DFMDDKRVYI MDVYNRHIYP GDRFAKQAIR RKVELEWGTE DDEYLDKVER NIKKSLQEHL PDVVVYNAGT DILEGDRLGG LSISPAGIVK RDELVFRMVR GRRVPILMVT SGGYQKRTAR IIADSILNLF GLGLIGPESP SVSAQNSDTP LLPPAVP.
This gene encodes a class IV histone deacetylase. The encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10. Alternative splicing results in multiple transcript variants.
Specifications
| HDAC11 | |
| Recombinant Monoclonal | |
| 0.9 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q91WA3, Q96DB2 | |
| HDAC11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-347 of human HDAC11 (NP_079103.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9I4N2 | |
| Unconjugated | |
| HDAC11 | |
| FLJ22237; HD11; HDAC11; Histone deacetylase 11 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 232232, 297453, 79885 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.