missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA511250
This item is not returnable.
View return policy
Description
HDAC11 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a class IV histone deacetylase. The encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10. Alternative splicing results in multiple transcript variants.
Specifications
| HDAC11 | |
| Polyclonal | |
| Unconjugated | |
| HDAC11 | |
| FLJ22237; HD11; HDAC11; Histone deacetylase 11 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 232232, 79885 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q91WA3, Q96DB2 | |
| HDAC11 | |
| KLH conjugated synthetic peptide between 313-345 amino acids from the C-terminal region of human HDAC11. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction