missing translation for 'onlineSavingsMsg'
Learn More

HDAC10 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18407222 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407222 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407222 Supplier Novus Biologicals Supplier No. NB600223MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HDAC10 Polyclonal antibody specifically detects HDAC10 in Human samples. It is validated for Western Blot, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC10
Applications Western Blot, Simple Western
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Simple Western
Formulation 50mM Sodium Borate
Gene Alias DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of human HDAC 10.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, Epigenetics, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 83933
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.