missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HDAC10 Polyclonal antibody specifically detects HDAC10 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | HDAC10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to a portion of human HDAC 10. |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?