missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC10 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-223CL1
This item is not returnable.
View return policy
Description
HDAC10 Polyclonal antibody specifically detects HDAC10 in Human samples. It is validated for Western Blot,Simple Western
Specifications
| HDAC10 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western | |
| CoraFluor 1 | |
| DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722 | |
| Synthetic peptide corresponding to a portion of human HDAC 10. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, Epigenetics, Transcription Factors and Regulators | |
| 83933 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction