missing translation for 'onlineSavingsMsg'
Learn More

HDAC1 Mouse anti-Human, Mouse, Rat, CoraLite 555, Clone: 4C4G7, Proteintech

Mouse Monoclonal Antibody

£135.00 - £375.00

Specifications

Antigen HDAC1
Clone 4C4G7
Applications Immunofluorescence
Classification Monoclonal
Conjugate CoraLite 555
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17731359
View Documents
Proteintech
CL555-66085-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17629906
View Documents
Proteintech
CL555-66085-100UL
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HDAC1 Monoclonal antibody apecifically detetects HDAC1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.
Specifications

Specifications

HDAC1
Immunofluorescence
CoraLite 555
Mouse
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
3065
HDAC1 fusion protein Ag19128
Primary
Store at -20°C. Avoid exposure to light.
4C4G7
Monoclonal
Liquid
Human, Mouse, Rat
BC000301
HDAC1
IgG1
Protein G purification
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.