missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC1 Monoclonal Antibody (CL0510)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA534985
This item is not returnable.
View return policy
Description
Immunogen sequence: RIACEEEFSD SEEEGEGGRK NSSNFKKAKR VKTEDEKEKD PEEKKEVTEE EKTKEEKPEA KGVKE Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.
Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. HDAC1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis.
Specifications
| HDAC1 | |
| Monoclonal | |
| Unconjugated | |
| HDAC1 | |
| DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 297893, 3065, 433759 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL0510 | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O09106, Q13547, Q4QQW4 | |
| HDAC1 | |
| Recombinant protein fragment of Human HDAC1 (Product #RP-95336) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction