missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC1 Monoclonal Antibody (1061CT1.3.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537620
This item is not returnable.
View return policy
Description
HDAC1 Monoclonal Antibody for Western Blot
Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. HDAC1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis.
Specifications
| HDAC1 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q13547 | |
| HDAC1 | |
| purified His-tagged HDAC1 protein. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| Western Blot | |
| 1061CT1.3.1 | |
| Unconjugated | |
| HDAC1 | |
| DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 | |
| Mouse | |
| Euglobulin precipitation | |
| RUO | |
| 3065 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction