missing translation for 'onlineSavingsMsg'
Learn More

MRS1845, MedChemExpress

Product Code. 16400207
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16400207

Brand: Medchem Express HY103310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRS1845 is a selective store-operated calcium (SOC) channel inhibitor with an IC50 of 1.7 μM. MRS1845 is an ORAI1 inhibitor.

MRS1845 is a selective store-operated calcium (SOC) channel inhibitor with an IC50 of 1.7 μM. MRS1845 is an ORAI1 inhibitor.

Specifications

Chemical Name or Material MRS1845
CAS 544478-19-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H22N2O6
Quantity 50 mg
Solubility Information DMSO : ≥ 83.33 mg/mL (209.16 mM)
SMILES O=C(C1=C(C)N(CC#C)C(C)=C(C(OC)=O)C1C2=CC=CC([N+]([O-])=O)=C2)OCC
Molecular Weight (g/mol) 398.41
Formula Weight 398.41
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.