missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCV-NS3 Antibody (1B6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80122F
This item is not returnable.
View return policy
Description
HCV-NS3 Monoclonal antibody specifically detects HCV-NS3 in Virus samples. It is validated for Immunofluorescence
Specifications
| HCV-NS3 | |
| Monoclonal | |
| FITC | |
| Nonstructural Protein 3, NS3 protease, Q91RS4 | |
| Recombinant HCV-NS3 serine protease domain. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunofluorescence | |
| 1B6 | |
| PBS | |
| Mouse | |
| >95%, by SDS-PAGE | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction