missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCP5 Monoclonal Antibody (3G7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521178
This item is not returnable.
View return policy
Description
Peptide Sequence: LRMSEHRNEA LGNYLEMRLK SSFLRGLGSW KSNPLRLGGW TILLTLTMGQ GEPGGPQGDP WVPHELLLPS LCDSSHASSW GSGSITCAWR GGDSSSHPLV S.
Specifications
| HCP5 | |
| Monoclonal | |
| 0.49 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q6MZN7 | |
| HCP5 | |
| HCP5 (NP_006665, 3 a.a. ~ 103 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 3G7 | |
| Unconjugated | |
| HCP5 | |
| 6S2650E; D6S2650E; GPHRB; HCP5; HLA complex P5 (non-protein coding); P5-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10866 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction