missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&DSystems™ HCoV-HKU1 Spike Protein (YP_173238.1) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | SARS-CoV-2 Spike |
| Accession Number | YP_173238.1 |
| Gene ID (Entrez) | 43740568 |
| Gene Alias | 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | HCoV-HKU1 |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction