missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCN4 Antibody (SHG 1E5), Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
£169.00
Specifications
| Antigen | HCN4 |
|---|---|
| Clone | SHG 1E5 |
| Dilution | Western Blot 1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:100, Immunohistochemistry-Frozen 1:1000, Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
HCN4 Monoclonal specifically detects HCN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
| HCN4 | |
| Western Blot 1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:100, Immunohistochemistry-Frozen 1:1000, Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Q9Y3Q4 | |
| 10021 | |
| Synthetic peptide sequence corresponding to residues S H G S L L L P P A S S P P P P Q V P Q R R G T P P L T P G R L T Q D L K L of HCN4. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SHG 1E5 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human | |
| hyperpolarization activated cyclic nucleotide-gated cation channel 4, hyperpolarization activated cyclic nucleotide-gated potassium channel 4, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4, SSS2 | |
| HCN4 | |
| IgG1 | |
| Unpurified | |
| HCN4 (SHG 1E5) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title