missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCN4 Antibody (S114/10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12910MFV500
This item is not returnable.
View return policy
Description
HCN4 Monoclonal antibody specifically detects HCN4 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry
Specifications
| HCN4 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| Purified |
| Flow Cytometry | |
| S114/10 | |
| Flow Cytometry | |
| hyperpolarization activated cyclic nucleotide-gated cation channel 4, hyperpolarization activated cyclic nucleotide-gated potassium channel 4, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4, SSS2 | |
| Fusion protein amino acids 1019-1198 (c-terminus) of rat HCN4 | |
| 0.1 mL | |
| Neuroscience | |
| 10021 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction