missing translation for 'onlineSavingsMsg'
Learn More

HCN2 Antibody (S71-37), DyLight 405, Novus Biologicals™

Product Code. 18424584 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424584 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424584 Supplier Novus Biologicals Supplier No. NBP212895V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HCN2 Monoclonal antibody specifically detects HCN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCN2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone S71-37
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias BCNG2, BCNG-2, brain cyclic nucleotide gated channel 2, brain cyclic nucleotide-gated channel 2, HAC-1, hyperpolarization activated cyclic nucleotide-gated potassium channel 2, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 2
Host Species Mouse
Immunogen Fusion protein amino acids 761-863 of rat HCN2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 610
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.