missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00
Specifications
| Antigen | HCN2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18793294
|
Novus Biologicals
NBP2-22518-0.025mg |
0.025 mg | |||||||
Description
HCN2 Polyclonal specifically detects HCN2 in Rat samples. It is validated for Western Blot.Specifications
| HCN2 | |
| Western Blot 1:500 | |
| Polyclonal | |
| Rabbit | |
| Rat | |
| BCNG2, BCNG-2, brain cyclic nucleotide gated channel 2, brain cyclic nucleotide-gated channel 2, HAC-1, hyperpolarization activated cyclic nucleotide-gated potassium channel 2, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 2 | |
| HCN2 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 1 mg/mL BSA with 0.05% Sodium Azide | |
| 610 | |
| Synthetic Peptide: G(874) L D P Q D S A R S R L S S N L(889) | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title