Learn More
Invitrogen™ HCN1 Monoclonal Antibody (RTQ 7C3), eBioscience™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ 14-5691-82
Product Code. 16586910
  • £440.00 / 100µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Description: This RTQ 7C3 monoclonal antibody recognizes mouse and rat HCN1. Clone RTQ 7C3 detects a ~110 kDa band corresponding to HCN1 by Western Blot. Applications Reported: This RTQ 7C3 antibody has been reported for use in western blot. Applications Tested: This RTQ 7C3 antibody has been tested by western blot of mouse and rat tissue lysates. This may be used at less than or equal to 1 μg/mL.
HCN1 includes Hyperpolarization-activated cation channels of the HCN gene family, which contribute to spontaneous rhythmic activity in both heart and brain. HCN1 is a member of a family of pacemaker channels activated by hyperpolarization and regulated by cyclic nucleotides. HCN1 and HCN2 play an important role for motor learning and neuronal integration by cerebellar Purkinje cells as well as, shaping autonomous activity of single neurons and the periodicity of network oscillations. The HCN1 expression is highly enriched in cerebral cortex, hippocampus, cerebellum, and facial motor nucleus. HCN2 is highly abundant in mamillary bodies, pontine nucleus, ventral cochlear nucleus, and nucleus of the trapezoid body. These variations in regional specificity of HCN channels could generate important differences in neuronal pacemaker activity across brain systems.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
HCN1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O88704, Q9JKB0
HCN1
aa 650 - 685 of rat HCN1.
100 μg
Primary
Mouse, Rat
Antibody
IgG1 κ
Western Blot
RTQ 7C3
Unconjugated
HCN1
Bcng1; BCNG-1; Brain cyclic nucleotide-gated channel 1; C630013B14Rik; EIEE24; Hac2; HAC-2; Hcn1; hyperpolarization activated cyclic nucleotide gated potassium channel 1; hyperpolarization-activated cation channel 2; hyperpolarization-activated cyclic nucleotide-gated potassium channel 1; hyperpolarization-activated, cyclic nucleotide-gated K+ 1; potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
Rat
Affinity chromatography
RUO
15165, 84390
4°C, do not freeze
Liquid
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications
Promotions
Promotions
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis