missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ HCM™ Hepatocyte Culture Medium BulletKit™

Product Code. 11685450
Change view
Click to view available options
Medium: 500 mL, Transferrin: 0.50 mL, Ascorbic Acid: 0.50 mL, HEGF: 0.50 mL, Insulin, 0.50 mL, Hydrocortisone: 0.50 mL, BSA (fatty acid free), 10.5 mL, GA-1000: 0.50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11685450 Medium: 500 mL, Transferrin: 0.50 mL, Ascorbic Acid: 0.50 mL, HEGF: 0.50 mL, Insulin, 0.50 mL, Hydrocortisone: 0.50 mL, BSA (fatty acid free), 10.5 mL, GA-1000: 0.50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11685450 Supplier Lonza™ Supplier No. CC3198

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Complete kit, basal medium and supplements, to support growth of primary hepatocytes.

  • Culture system containing HBM™ Basal Medium (CC-3199), and; HCM SingleQuots™ Supplement Pack (CC-4182).
  • Developed to support growth of primary hepatocytes.
  • Highly reproducible maintenance medium for primary hepatocytes from all species.
  • Optimized for hepatocyte cell health and compatible with all hepatocyte species.
  • The final medium is serum-free.
  • Includes the following:
    • 1 x HBM™ Basal Medium (CC-3199), 500 mL
    • 1 x HCM SingleQuots™ Supplement Pack (CC-4182) containing the following:
      • 1 x Transferrin, 0.50 mL
      • 1 x Ascorbic Acid, 0.50 mL
      • 1 x HEGF, 0.50 mL
      • 1 x Insulin, 0.50 mL
      • 1 x Hydrocortisone, 0.50 mL
      • 1 x BSA (fatty acid free), 10.5 mL
      • 1 x GA-1000, 0.50 mL
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hepatocyte Cells
Product Type Basal Medium and Supplements
Quantity Medium: 500 mL, Transferrin: 0.50 mL, Ascorbic Acid: 0.50 mL, HEGF: 0.50 mL, Insulin, 0.50 mL, Hydrocortisone: 0.50 mL, BSA (fatty acid free), 10.5 mL, GA-1000: 0.50 mL
pH 7.2 to 7.6
Color Clear
Content And Storage Basal Medium, Transferrin, Ascorbic Acid, HEGF, Insulin, Hydrocortisone, BSA (fatty acid free), GA-1000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.