Learn More
HCLS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25003-1-AP-150UL
 View more versions of this product
Product Code. 16873525
  • £349.00 / 150µL
Description
This gene encodes a deubiquitinating enzyme that is phosphorylated at the onset of mitosis and then dephosphorylated at the metaphase/anaphase transition. It can deubiquitinate H2A, one of two major ubiquitinated proteins of chromatin, in vitro and a mutant form of the protein was shown to block cell division. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
HCLS1
Polyclonal
Unconjugated
HCLS1
CTTNL, HCLS1, HS1, LckBP1, p75
Rabbit
Antigen Affinity Chromatography
RUO
3059
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14317
HCLS1
HCLS1 Fusion Protein Ag8645
150 μL