missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCLS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102594
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HCLS1.
Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression. [UniProt].
Specifications
| HCLS1 | |
| Polyclonal | |
| Unconjugated | |
| HCLS1 | |
| AW213261; CTTNL; HCLS1; hematopoietic cell specific Lyn substrate 1; hematopoietic cell-specific Lyn substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15163, 288077, 3059 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14317, P49710 | |
| HCLS1 | |
| A synthesized peptide derived from human HCLS1(Accession P14317), corresponding to amino acid residues L173-K223. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction