missing translation for 'onlineSavingsMsg'
Learn More

Hck Rabbit anti-Human, Clone: S02-5C1, Novus Biologicals™

Product Code. 18386702 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386702 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386702 Supplier Novus Biologicals Supplier No. NBP31495725UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Hck Monoclonal antibody specifically detects Hck in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hck
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S02-5C1
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias EC 2.7.10, EC 2.7.10.2, hemopoietic cell kinaseJTK9, p59-HCK/p60-HCK, tyrosine-protein kinase HCK
Host Species Rabbit
Immunogen A synthetic peptide of human Hck (Uniprot#: P08631)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3055
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.