Learn More
Eurofins DiscoverX Hck Protein, Activated
Description
Hematopoietic cell kinase (HCK) is a member of Src protein-tyrosine kinase family and expressed primary in granulocytes, monocytes and macrophages. Active HCK phosphorylates STAT5B on Tyr699, which represents an essential step in Signal transducer and activator of transcription 5 (STAT5B) stimulation. The binding of SH3 ligands, such as the Nef protein of HIV-1, further augments Hck activity suggesting a new mechanism for regulating tyrosine kinase activity.
GenBank NM_002110
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 5 mM β-glycerophosphate, 1 mM Na3VO4. Frozen solution. |
| Molecular Weight (g/mol) | 34 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥63% |
| Protein | Hck, activated |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.