missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HCK, GST Tag Recombinant Protein

Product Code. 11330872
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
1mg
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11330872

Brand: Invitrogen™ PV6128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Hematopoietic cell kinase (HCK) is a SRC-related kinase expressed in lymphoid and hematopoietic cells and may play a role in some forms of leukemia. (HCK Alias: DKFZp686D0638). HCK is a protein-tyrosine kinase that is predominantly expressed in hemopoietic cell types. The encoded protein may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Alternate translation initiation site usage, including a non-AUG (CUG) codon, results in the production of two different isoforms, that have different subcellular localization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08631
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.02% NP-40, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 3055
Molecular Weight (g/mol) 84.8 kDa
Name Human HCK, GST Tag
Purification Method Purified
Quantity 10 μg
Gene Alias AI849071; B cell/myeloid kinase; B-cell/myeloid kinase; BMK; HCK; HCK proto-oncogene, Src family tyrosine kinase; Hctk; Hematopoietic cell kinase; hemopoietic cell kinase; hemopoietic cell tyrosine kinase; JTK9; p56Hck; p56-HCK; p56-HCK/p59-HCK; p59Hck; p59-HCK/p60-HCK; p61Hck; tyrosine-protein kinase HCK
Common Name HCK
KinaseFamily SRC Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Protein Family Kinases & Inhibitors
Form Liquid
Protein Subtype Cytoplasmic Tyrosine Kinases
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.