missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Hck Polyclonal antibody specifically detects Hck in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | Hck |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | EC 2.7.10, EC 2.7.10.2, hemopoietic cell kinaseJTK9, p59-HCK/p60-HCK, tyrosine-protein kinase HCK |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 350-400 on human Hematopoietic cell kinase/ Tyrosine-protein kinase HCK. |
| Purification Method | Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?