missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hCG Monoclonal Antibody (3002)

Description
The observed molecular weight of alpha-hCG and beta-hCG are approximately 20-22 kD & 27-32 kD respectively. The molecular weight is higher than the predicted molecular weight due to post translational modification. MIH9801 was formerly sold as a Seradyn product.
Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregnancy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones including pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. The alpha subunit is common to all the members of the family within a given species. The beta subunits are distinct in each of the horomones and confer the receptor and biological specificity.
Specifications
Specifications
| Antigen | hCG |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | 3002 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CGB3 |
| Gene Accession No. | P01215, P0DN86 |
| Gene Alias | anterior pituitary glycoprotein hormones common subunit alpha; CGA; CG-alpha; CGB; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin alpha chain; choriogonadotropin subunit beta; Choriogonadotropin subunit beta 3; chorionic gonadotrophin chain beta; chorionic gonadotrophin subunit alpha; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta 5 subunit; chorionic gonadotropin beta 8 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit; chorionic gonadotropin beta subunit 3; chorionic gonadotropin beta subunit 5; chorionic gonadotropin beta subunit 8; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, alpha polypeptide; chorionic gonadotropin, beta polypeptide; chorionic gonadotropin, beta polypeptide 5; chorionic gonadotropin, beta polypeptide 8; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin alpha chain; FSHA; FSH-alpha; Glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; GPHa; GPHA1; HCG; hCGB; human chorionic gonadotropin; LHA; LSH-alpha; luteinizing hormone alpha chain; Lutropin alpha chain; thyroid-stimulating hormone alpha chain; Thyrotropin alpha chain; TSHA; TSH-alpha |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?