missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCG Holo Mouse anti-Human, Clone: HCGab/52, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14427.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against intact-HCG.
- High specificity
- High reactivity
Specifications
| HCG Holo | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CG-ALPHA/FSHA/GPHA1/GPHa/HCG/LHA/TSHA | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 1081/1082 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| HCGab/52 | |
| Mouse monoclonal antibody raised against intact-HCG. | |
| In 10 mM PBS (0.05% BSA and 0.05% azide). | |
| CGA | |
| Native purified hCG protein. | |
| 100 μg | |
| Primary | |
| Reacts only with the intact-HCG and not with either free alpha- or free beta-chain of HCG. | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction