missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hCG beta Monoclonal Antibody (INN-hCG-68)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA182291
This item is not returnable.
View return policy
Description
Clone INN-hCG-68 recognizes hCG (Beta 7 epitope) under non-reducing conditions. MA1-82291 recognized free human chorionic gonadotrophin beta (hCG beta), but does not bind to holo-hCG, holo-hLH or free hLH beta.
hCG beta is the beta chain of hCG (Human chorionic gonadotropin), a 40 kDa glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. hCG is made up of alpha and beta subunits that are non-covalently associated. The alpha subunit is similar to those found on the pituitary hormones FSH, LH ans TSH. However, the beta subunit is different in FSH, LH and TSH and are involved in conferring biological specificity. Maintenance of the fetus in the first trimester of pregnancy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium. hCG is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and Beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). Diseases associated with Hcg dysfunction include gestational trophoblastic tumor and hydatidiform mole, recurrent 1.
Specifications
| hCG beta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P0DN86 | |
| CGB3 | |
| Proprietary Immunogen. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Radioimmune Assays (RIA), Western Blot | |
| INN-hCG-68 | |
| Unconjugated | |
| CGB3 | |
| CGB; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta; choriogonadotropin subunit beta 3; chorionic gonadotrophin chain beta; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit; chorionic gonadotropin beta subunit 3; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide; FSH-alpha; hCGB; human chorionic gonadotropin; LSH-alpha; TSH-alpha | |
| Mouse | |
| Protein A | |
| RUO | |
| 1082 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction