missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hCG beta-7 Monoclonal Antibody (6F12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524507
This item is not returnable.
View return policy
Description
Peptide Sequence: GYCPTMTRVL QGVLPALPQV VCNYRDVRFE SIRLPGCPRG VNPVVSYAVA LSCQCALCRR STTDCGGPKD HPLTCDDPRF QASSSSKAPP PSLPSPSRLP GPSDTPILPQ.
This gene is a member of the glycoprotein hormone beta chain family and encodes the beta 7 subunit of chorionic gonadotropin (CG). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. CG is produced by the trophoblastic cells of the placenta and stimulates the ovaries to synthesize the steroids that are essential for the maintenance of pregnancy. The beta subunit of CG is encoded by 6 genes which are arranged in tandem and inverted pairs on chromosome 19q13.3 and contiguous with the luteinizing hormone beta subunit gene.
Specifications
| hCG beta-7 | |
| Monoclonal | |
| Unconjugated | |
| CGB7 | |
| CGB6; CGB7; CG-beta-a; Choriogonadotropin subunit beta 7; chorionic gonadotropin beta 7 subunit; chorionic gonadotropin beta subunit 7; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 7; human chorionic gonadotropin | |
| Mouse | |
| Protein A | |
| RUO | |
| 94027 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 6F12 | |
| PBS with no preservative; pH 7.4 | |
| P0DN87 | |
| CGB7 | |
| CGB7 (NP_149133, 56 a.a. ~ 165 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction