missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ hCG beta-1 Recombinant Rabbit Monoclonal Antibody (K03_1J44)

Product Code. 30830043
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830043 50 μL
30829213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830043 Supplier Invitrogen™ Supplier No. MA564559
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

hCG beta-1 Recombinant Monoclonal Antibody for IHC (P)

HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and Beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).
TRUSTED_SUSTAINABILITY

Specifications

Antigen hCG beta-1
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K03_1J44
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CGB1
Gene Accession No. A6NKQ9
Gene Alias CGB1; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta variant 1; chorionic gonadotropin beta subunit 1; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 1; hCGB; human chorionic gonadotropin
Gene Symbols CGB1
Host Species Rabbit
Immunogen Recombinant human CGB1 protein.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114335
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.