missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hCG beta-1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£155.00 - £386.00
Specifications
| Antigen | hCG beta-1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17783314
|
Invitrogen™
PA5121920 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17773314
![]() |
Invitrogen™
PA5121919 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and Beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).Specifications
| hCG beta-1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| A6NKQ9 | |
| 114335 | |
| Tyr55-Pro153 (Accession A6NKQ9), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CGB1 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| CGB1; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta variant 1; chorionic gonadotropin beta subunit 1; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 1; hCGB; human chorionic gonadotropin | |
| CGB1 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title