missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hCG beta-1 Monoclonal Antibody (INN-hCG-2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516740
This item is not returnable.
View return policy
Description
Mouse anti hCG antibody, clone INN-hCG-2 recognizes the beta 1 epitope on human chorionic gonadotrophin (hCG).Chorionic gonadotrophin is a member of the glycoprotein hormone family that is synthesized by the placenta throughout pregnancy.
HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and Beta subunits. The alpha subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH).
Specifications
| hCG beta-1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| A6NKQ9 | |
| CGB1 | |
| hCG. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
| INN-hCG-2 | |
| Unconjugated | |
| CGB1 | |
| CGB1; CGB3; CGB5; CGB7; CGB8; CG-beta; Choriogonadotropin subunit beta variant 1; chorionic gonadotropin beta subunit 1; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 1; hCGB; human chorionic gonadotropin | |
| Mouse | |
| Protein A | |
| RUO | |
| 114335 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction