missing translation for 'onlineSavingsMsg'
Learn More

hCG Antibody, DyLight 550, Novus Biologicals™

Product Code. 18367673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367673 0.1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367673 Supplier Novus Biologicals Supplier No. NBP311024R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

hCG Polyclonal antibody specifically detects hCG in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen hCG
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen Human TSH (intact)
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.