Learn More
Invitrogen™ hCG alpha Human ELISA Kit
Sandwich ELISA Kit
£560.00 - £2320.00
Specifications
Accession Number | P01215 |
---|---|
Assay Sensitivity | 50 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15484943
|
Invitrogen™
EHCG |
96 Tests |
£560.00
1 set |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
17125399
|
Invitrogen™
EHCGX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
The Human Chorionic Gonadotropin (hCG) ELISA quantitates Hu CG in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CG. Principle of the method The Human CG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.Specifications
P01215 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
1081 | |
<12% | |
HRP | |
Apoptosis | |
RP11-156F23.1, CG-ALPHA, FSHA, GPHA1, GPHa, HCG, LHA, TSHA | |
1 hr. 20 min. |
50 pg/mL | |
Biotin | |
Other Proteins | |
Human | |
Colorimetric Microplate Reader | |
CG-ALPHA,FSHA,GPHA1,GPHa,HCG,LHA,TSHA | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |