missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCFC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102495
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HCFC1.
Tumor necrosis factor (TNF) receptor associated factors (TRAFs) were initially discovered as adaptor proteins that link the TNF receptor superfamily to signaling pathways and are thus important regulators of cell death and cellular response to stress. TRAF proteins share a homology region that allows them to bind to cell receptors and other TRAF proteins, causing the activation of different signal cascades depending on the TRAFs involved. For example, TRAF2 and TRAF3 directly bind to the CD40, a NF receptor superfamily member involved in inducing B cell immunity, and are critical for NF-kappa-B activation in mouse B lymphocytes. TRAF2 along with TRAF6 has also been shown to be required for CD40 signaling in nonhemopoietic cells. TRAF2 also interacts with the TRFR superfamily member lymphotoxin-beta receptor (LTbetaR) in association with TRAF3 and the apoptosis inhibitors cIAP1 and Smac.
Specifications
| HCFC1 | |
| Polyclonal | |
| Unconjugated | |
| HCFC1 | |
| AW060991; C1 factor; CFF; HCF; HCF C-terminal chain 1; HCF C-terminal chain 2; HCF C-terminal chain 3; HCF C-terminal chain 4; HCF C-terminal chain 5; HCF C-terminal chain 6; HCF N-terminal chain 1; HCF N-terminal chain 2; HCF N-terminal chain 3; HCF N-terminal chain 4; HCF N-terminal chain 5; HCF N-terminal chain 6; HCF1; HCF-1; hcfc1; HFC1; host cell factor 1; host cell factor C1; host cell factor C1 (VP16-accessory protein); LOW QUALITY PROTEIN: host cell factor 1; MRX3; PPP1R89; protein phosphatase 1, regulatory subunit 89; RGD1563804; VCAF; VP16 accessory protein; VP16-accessory protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15161, 3054, 363519 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51610, Q61191 | |
| HCFC1 | |
| A synthesized peptide derived from human HCFC1(Accession P51610), corresponding to amino acid residues R200-V250. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction