missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCE Recombinant Rabbit Monoclonal Antibody (K01_2M60)

Description
HCE Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.
Specifications
Specifications
| Antigen | HCE |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | K01_2M60 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | RNGTT |
| Gene Accession No. | O60942 |
| Gene Alias | AU020997; CAP1A; GTase; GTP--RNA guanylyltransferase; hCAP; HCAP1; HCE; HCE1; MCE1; mRNA 5'-triphosphatase; mRNA guanylyltransferase; mRNA-capping enzyme; Polynucleotide 5'-triphosphatase; RNA guanylyltransferase and 5'-phosphatase; Rngtt; RP3-488C13.1; TPase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?