missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521684
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (95%), Rat (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.
Specifications
| HCE | |
| Polyclonal | |
| Unconjugated | |
| RNGTT | |
| AU020997; CAP1A; GTase; GTP--RNA guanylyltransferase; hCAP; HCAP1; HCE; HCE1; MCE1; mRNA 5'-triphosphatase; mRNA guanylyltransferase; mRNA-capping enzyme; Polynucleotide 5'-triphosphatase; RNA guanylyltransferase and 5'-phosphatase; Rngtt; RP3-488C13.1; TPase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8732 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.54 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O60942 | |
| RNGTT | |
| Recombinant protein encompassing a sequence within the center region of human HCE. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction