missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCCS Polyclonal Antibody

HCCS Polyclonal Antibody
Brand: Thermo Scientific PA5106551
This item is not returnable.
View return policy
Description
HCCS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| HCCS | |
| Polyclonal | |
| Unconjugated | |
| HCCS | |
| CCHL; cytochrome c heme-lyase; cytochrome c-type heme lyase; HCCS; holocytochrome c synthase; holocytochrome c synthetase; holocytochrome c-type synthase; LSDMCA1; MCOPS7; microphthalamia with linear skin defects; MLS; RGD1563855 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15159, 3052, 317444 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P53701, P53702 | |
| HCCS | |
| A synthesized peptide derived from human HCCS(Accession P53701), corresponding to amino acid residues L84-G134. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction