missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HCC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590681
This item is not returnable.
View return policy
Description
Immunogen sequence: ATQCFQLSNM FNPQTEEEVG WDTEIKDDVI EECNKHGGVI HIYVDKNSAQ GNVYVKCPSI AAAIAAVNAL HGRWFAGKMI TAAYVPLPTY HNLFPDSMTA TQLLVPSRR; Positive Samples: BxPC-3, U-251MG, 293T, LO2, Mouse brain, Mouse heart; Cellular Location: Nucleus speckle.
The protein encoded by this gene is an RNA binding protein and possible splicing factor. The encoded protein is found in the nucleus, where it colocalizes with core spliceosomal proteins. Studies of a mouse protein with high sequence similarity to this protein suggest that this protein may act as a transcriptional coactivator for JUN/AP-1 and estrogen receptors. Multiple transcript variants encoding different isoforms have been observed for this gene.
Specifications
| HCC1 | |
| Polyclonal | |
| Unconjugated | |
| RBM39 | |
| 1500012C14Rik; 2310040E03Rik; B330012G18Rik; C79248; CAPER; CAPER alpha; CAPERalpha; Coactivator of activating protein 1 and estrogen receptors; coactivator of activating protein-1 and estrogen receptors; coactivator of AP-1 and ERs; FSAP59; functional spliceosome-associated protein 59; HCC1; Hepatocellular carcinoma protein 1; R75070; RBM39; RNA binding motif protein 39; RNA-binding motif protein 39; RNA-binding protein 39; RNA-binding region (RNP1, RRM) containing 2; RNA-binding region-containing 2; RNA-binding region-containing protein 2; Rnp1; Rnpc2; Rrm; Splicing factor HCC1; transcription coactivator CAPER | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170791, 362251, 9584 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.66 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q14498, Q8VH51 | |
| RBM39 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-508 of human RBM39 (NP_0012295281). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction