missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCC1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-333CL1
This item is not returnable.
View return policy
Description
HCC1 Polyclonal antibody specifically detects HCC1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| HCC1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| CAPER, CC1.3, coactivator of activating protein-1 and estrogen receptors, DKFZp781C0423, FLJ44170, fSAP59, functional spliceosome-associated protein 59, HCC1CAPERalpha, Hepatocellular carcinoma protein 1, RNA binding motif protein 39, RNA-binding motif protein 39, RNA-binding protein 39, RNA-binding region-containing protein 2, RNPC2, RRM) containing 2, splicing factor CC1.3, Splicing factor HCC1 | |
| The immunogen this antibody was made to, maps to a region between residues 325-375 of human Caper, Coactivator of activating protein-1 and estrogen receptors using the SwissProt entry Q14498 (GeneID 9584). | |
| 0.1 mL | |
| Cellular Markers | |
| 9584 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction